Merck
  • Leveraging the Coffee Ring Effect for a Defect-Free Electroformation of Giant Unilamellar Vesicles.
Leveraging the Coffee Ring Effect for a Defect-Free Electroformation of Giant Unilamellar Vesicles.
Langmuir : the ACS journal of surfaces and colloids (2019-11-21)
Eric Oropeza-Guzman, Maricarmen Ríos-Ramírez, Jesús Carlos Ruiz-Suárez
ABSTRACT
We took advantage of the microflow hydrodynamics in the evaporation of sessile droplets to increase the height uniformity of thin lipid films for the subsequent electroformation of defect-free giant unilamellar vesicles (GUV). By serially casting progressively larger liposome suspension droplets on the same spot of an indium-tin-oxide (ITO) electrode, we managed to leverage the coffee ring effect (CRE) in the evaporation of each droplet to generate a smeared multilayer film of uniform thickness. This multidroplet technique of lipid film formation outperformed the traditional single-droplet deposition, improving the final quality of electroformed GUV samples. The proposed film formation technique constitutes a solvent-free method that results in a dramatic reduction (∼20×) in the appearance of undesirable structures like nonspherical (NSV), multilamellar (MLV), and multivesicular (MVV) vesicles.
Product Description
Avanti
14:0-12:0 NBD PC, Avanti Research - A Croda Brand 810123P, powder
Pricing and availability is not currently available.