Merck
  • Ultrasound-induced emission color and transmittance changes of organogel based on "trans-to-cis" isomerization.
Ultrasound-induced emission color and transmittance changes of organogel based on "trans-to-cis" isomerization.
Ultrasonics sonochemistry (2019-08-28)
Lijun Geng, Xudong Yu, Yanqiu Wang, Yajuan Li, Fengjuan Shen, Jujie Ren
ABSTRACT
Herein, instant and precise control of fluorescent emission color and transmittance could be carried out by ultrasound-promoted gel-to-gel transition of naphthalimide derivatives containing CN unit. It is proved that ultrasound triggered an irreversible and efficient configuration transformation of N1 from "trans to cis" form in gel state, which is stabilized by intermolecular hydrogen bonding interaction and not observed in the solution state.
Sigma-Aldrich
2-Hydroxy-4-methoxybenzaldehyde, 98%
Sigma-Aldrich
Cholesteryl chloroformate, 95%