Merck
  • Analysis of CCDC6 as a novel biomarker for the clinical use of PARP1 inhibitors in malignant pleural mesothelioma.
Analysis of CCDC6 as a novel biomarker for the clinical use of PARP1 inhibitors in malignant pleural mesothelioma.
Lung cancer (Amsterdam, Netherlands) (2019-08-27)
Francesco Morra, Francesco Merolla, Debora D'Abbiero, Gennaro Ilardi, Severo Campione, Roberto Monaco, Gianluca Guggino, Francesca Ambrosio, Stefania Staibano, Aniello Cerrato, Roberta Visconti, Angela Celetti
ABSTRACT
CCDC6 (coiled-coil domain containing 6) is a player of the HR response to DNA damage and has been predicted to interact with BAP1, another HR-DNA repair gene highly mutated in Malignant Pleural Mesothelioma (MPM), an aggressive cancer with poor prognosis. CCDC6 levels are modulated by the deubiquitinase USP7, and CCDC6 defects have been reported in several tumors determining PARP-inhibitors sensitivity. Our aim was to investigate the functional role of CCDC6 in MPM carcinogenesis and response to PARP-inhibitors. The interaction between CCDC6 and BAP1 was confirmed in MPM cells, by co-immunoprecipitation. Upon USP7 inhibition, that induces CCDC6 degradation, the ability to repair the DSBs and the sensitivity to PARP inhibitors, was explored by HR reporter and by cells viability assays, respectively. A TMA including 34 MPM cores was immunostained for CCDC6, USP7 and BAP1 and the results correlated by statistical analysis. MPM cells depleted of CCDC6 showed defects in DSBs repair and sensitivity to PARP inhibitors. The silencing of CCDC6 when combined with the overexpression of BAP1-mutant (Δ221-238) enhanced the HR-DNA repair defects and the PARP inhibitors sensitivity. In the TMA of MPM primary samples, the staining of CCDC6 and of its de-ubiquitinase USP7 showed a significant correlation in the tested primary samples (p = 0.01). CCDC6 was barely detected in 30% of the tumors that also carried BAP1 defects. The combination of CCDC6 and BAP1 staining may indicate therapeutic options for DDR targeting, acting in synergism with cisplatinum.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCDC6