Merck
  • Tanshinone IIA attenuates demyelination and promotes remyelination in A. cantonensis-infected BALB/c mice.
Tanshinone IIA attenuates demyelination and promotes remyelination in A. cantonensis-infected BALB/c mice.
International journal of biological sciences (2019-10-09)
Ying Feng, Feng Feng, Cunjing Zheng, Zongpu Zhou, Meihua Jiang, Zhen Liu, Fukang Xie, Xi Sun, Zhongdao Wu
ABSTRACT
Angiostrongylus cantonensis infection can cause demyelination in the central nervous system, and there is no effective treatment. We used dexamethasone, Tanshinone IIA (TSIIA) and Cryptotanshinone(Two traditional Chinese medicine monomers) in combination with albendazole (AB, a standard anti-helminthic compound) to observe their therapeutic effect on demyelination in A. cantonensis-infected mice. Luxol fast blue staining and electron microscope of myelin sheath, Oligodendrocyte (OL) number and myelin basic protein (MBP) expression in brain was detected in above groups. TSIIA+AB facilitated OL proliferation and significantly increased both myelin sheath thickness and the population of small-diameter axons. In addition, TSIIA treatment inhibited the expression of inflammation-related factors (interleukin [IL]-6, IL-1β, tumor necrosis factor [TNF]-α, inducible nitric oxide synthase [iNOS]) rather than inhibiting eosinophil infiltration in brain. TSIIA also decreased microglial activation and shifted their phenotype from M1 to M2. Taken together, these results provide evidence that TSIIA combined with AB may be an effective treatment for demyelination caused by A. cantonensis infection and other demyelinating diseases.
Sigma-Aldrich
Monoclonal Anti-Iba1 antibody produced in mouse, clone GT10312, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Oligodendrocyte Marker O4 antibody produced in mouse, clone O4, purified immunoglobulin, lyophilized powder