Merck
  • Using T. brucei as a biological epitope-display platform to elicit specific antibody responses.
Using T. brucei as a biological epitope-display platform to elicit specific antibody responses.
Journal of immunological methods (2010-08-31)
Pete Stavropoulos, F Nina Papavasiliou
ABSTRACT
The African trypanosome (Trypanosoma brucei) is transmitted by the bite of the tsetse vector to the mammalian bloodstream where it exists as a completely extracellular parasite. As a result of this exposure, the parasite elicits a robust immune response that is almost exclusively antibody mediated, and is extremely specific to the trypanosome coat displayed on the surface. This coat is comprised of ~11 million copies of a single gpi-linked molecule (the variable surface glycoprotein or VSG) and can therefore be used as a powerful platform for the immunogenic display of antigenic determinants. Here we describe a method to display repetitive, ordered arrays of linear epitopes on the surface of T. brucei and to then use the engineered organisms to generate specific anti-epitope antibody responses, upon injection into mice. This method offers an alternative approach to generating anti-peptide antibodies, and could be a useful option in cases where more traditional methods have failed.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-FITC antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.