Merck
  • High-throughput screening for human lysosomal beta-N-Acetyl hexosaminidase inhibitors acting as pharmacological chaperones.
High-throughput screening for human lysosomal beta-N-Acetyl hexosaminidase inhibitors acting as pharmacological chaperones.
Chemistry & biology (2007-02-24)
Michael B Tropak, Jan E Blanchard, Stephen G Withers, Eric D Brown, Don Mahuran
ABSTRACT
The adult forms of Tay-Sachs and Sandhoff diseases result when the activity of beta-hexosaminidase A (Hex) falls below approximately 10% of normal due to decreased transport of the destabilized mutant enzyme to the lysosome. Carbohydrate-based competitive inhibitors of Hex act as pharmacological chaperones (PC) in patient cells, facilitating exit of the enzyme from the endoplasmic reticulum, thereby increasing the mutant Hex protein and activity levels in the lysosome 3- to 6-fold. To identify drug-like PC candidates, we developed a fluorescence-based real-time enzyme assay and screened the Maybridge library of 50,000 compounds for inhibitors of purified Hex. Three structurally distinct micromolar competitive inhibitors, a bisnaphthalimide, nitro-indan-1-one, and pyrrolo[3,4-d]pyridazin-1-one were identified that specifically increased lysosomal Hex protein and activity levels in patient fibroblasts. These results validate screening for inhibitory compounds as an approach to identifying PCs.