Merck
  • Cyclooxygenase-2 stimulates production of amyloid beta-peptide in neuroblastoma x glioma hybrid NG108-15 cells.
Cyclooxygenase-2 stimulates production of amyloid beta-peptide in neuroblastoma x glioma hybrid NG108-15 cells.
Biochemical and biophysical research communications (2001-02-22)
K Kadoyama, Y Takahashi, H Higashida, T Tanabe, T Yoshimoto
ABSTRACT
Cyclooxygenase (COX) synthesizes bioactive prostaglandins from arachidonic acid, and there are COX-1 and COX-2 isoforms with distinct pathophysiological functions. Recent studies demonstrated that COX-2 expression was up-regulated in the brain of patients with Alzheimer's disease. We established mouse neuroblastoma x rat glioma hybrid NG108-15 cells stably expressing human COX-2. The COX-2-expressing cells showed 3- to 4-fold increases in both COX activity and prostaglandin E(2) production. The mRNA level of amyloid precursor protein (APP) was elevated by approximately 2-fold in the COX-2-expressing cells compared with mock-transfected cells. Amyloid beta-peptide and a secreted form of APP, both derived from APP by proteolysis was also increased. Interestingly, neurite outgrowth was stimulated in the COX-2-expressing cells with concomitant reduction of the cell proliferation rate. A selective COX-2 inhibitor (JTE-522) and a nonselective COX inhibitor (indomethacin) suppressed production of amyloid beta-peptide and a secreted form of APP by inhibition of APP mRNA level, suggesting that COX-2 plays important roles in the neurodegenerative processes of Alzheimer's disease.
Sigma-Aldrich
Indomethacin, A non-steroidal anti-inflammatory, cell permeable, antipyretic agent.
Pricing and availability is not currently available.