Merck
  • Interleukin-17 Drives Interstitial Entrapment of Tissue Lipoproteins in Experimental Psoriasis.
Interleukin-17 Drives Interstitial Entrapment of Tissue Lipoproteins in Experimental Psoriasis.
Cell metabolism (2018-11-13)
Li-Hao Huang, Bernd H Zinselmeyer, Chih-Hao Chang, Brian T Saunders, Andrew Elvington, Osamu Baba, Thomas J Broekelmann, Lina Qi, Joseph S Rueve, Melody A Swartz, Brian S Kim, Robert P Mecham, Helge Wiig, Michael J Thomas, Mary G Sorci-Thomas, Gwendalyn J Randolph
ABSTRACT
Lipoproteins trapped in arteries drive atherosclerosis. Extravascular low-density lipoprotein undergoes receptor uptake, whereas high-density lipoprotein (HDL) interacts with cells to acquire cholesterol and then recirculates to plasma. We developed photoactivatable apoA-I to understand how HDL passage through tissue is regulated. We focused on skin and arteries of healthy mice versus those with psoriasis, which carries cardiovascular risk in man. Our findings suggest that psoriasis-affected skin lesions program interleukin-17-producing T cells in draining lymph nodes to home to distal skin and later to arteries. There, these cells mediate thickening of the collagenous matrix, such that larger molecules including lipoproteins become entrapped. HDL transit was rescued by depleting CD4+ T cells, neutralizing interleukin-17, or inhibiting lysyl oxidase that crosslinks collagen. Experimental psoriasis also increased vascular stiffness and atherosclerosis via this common pathway. Thus, interleukin-17 can reduce lipoprotein trafficking and increase vascular stiffness by, at least in part, remodeling collagen.
Sigma-Aldrich
FTY720, ≥98% (HPLC)
Pricing and availability is not currently available.