- Production of Tissue-Engineered Small Intestine in Rats with Different Ages of Cell Donors.
Production of Tissue-Engineered Small Intestine in Rats with Different Ages of Cell Donors.
Tissue engineering. Part A (2018-10-05)
Yanchun Liu, Yijie Wang, Jason Chakroff, Jed Johnson, Aidan Farrell, Gail E Besner
PMID30284958
ABSTRACT
This study compared side-by-side the impact of donor age on the production of tissue-engineered small intestine (TESI). Each age represents a specific period of life: E18 for fetuses, 5-day-old pups for neonates, 21-day-old rats for weanlings, and 6-week-old rats for adults. The TESI produced was compared macroscopically and microscopically. The mechanism(s) contributing to the differences observed was explored by detecting proliferating cells in the TESI and by analyzing intestinal stem cell gene expression in donor cells. These data may provide valuable information for future application of TESI clinically.