Merck
  • Experimental Validation of Hydrogen Atom Transfer Gibbs Free Energy as a Predictor of Nitroaromatic Reduction Rate Constants.
Experimental Validation of Hydrogen Atom Transfer Gibbs Free Energy as a Predictor of Nitroaromatic Reduction Rate Constants.
Environmental science & technology (2019-05-01)
Jimmy Murillo-Gelvez, Kevin P Hickey, Dominic M Di Toro, Herbert E Allen, Richard F Carbonaro, Pei C Chiu
ABSTRACT
Nitroaromatic compounds (NACs) are a class of prevalent contaminants. Abiotic reduction is an important fate process that initiates NAC degradation in the environment. Many linear free energy relationship (LFER) models have been developed to predict NAC reduction rates. Almost all LFERs to date utilize experimental aqueous-phase one-electron reduction potential ( EH1) of NAC as a predictor, and thus, their utility is limited by the availability of EH1 data. A promising new approach that utilizes computed hydrogen atom transfer (HAT) Gibbs free energy instead of EH1 as a predictor was recently proposed. In this study, we evaluated the feasibility of HAT energy for predicting NAC reduction rate constants. Using dithionite-reduced quinones, we measured the second-order rate constants for the reduction of seven NACs by three hydroquinones of different protonation states. We computed the gas-phase energies for HAT and electron affinity (EA) of NACs and established HAT- and EA-based LFERs for six hydroquinone species. The results suggest that HAT energy is a reliable predictor of NAC reduction rate constants and is superior to EA. This is the first independent, experimental validation of HAT-based LFER, a new approach that enables rate prediction for a broad range of structurally diverse NACs based solely on molecular structures.