Merck
  • NLRP3 inflammasome activates interleukin-23/interleukin-17 axis during ischaemia-reperfusion injury in cerebral ischaemia in mice.
NLRP3 inflammasome activates interleukin-23/interleukin-17 axis during ischaemia-reperfusion injury in cerebral ischaemia in mice.
Life sciences (2019-04-20)
Haining Wang, Di Zhong, Hongping Chen, Jing Jin, Qingqing Liu, Guozhong Li
ABSTRACT
NLRP3 inflammasome has been reported associated with some inflammatory and autoimmune diseases. We previously researches showed that interleukin-23 (IL-23) and interleukin-17 (IL-17) aggravates the ischaemic injury of the brain tissue. However, it is poorly understood whether the NLPR3 inflammasome was involved in regulating and activating the IL-23/IL-17 axis in ischaemic stroke. We aimed to delineate whether the NLRP3 inflammasome signalling provokes the IL-23/IL-17 axis and interleukin-23 receptor (IL-23R) inducing the ischaemia-reperfusion injury of the brain in mice. The male C57/BL6 mice with experimental transient middle cerebral artery occlusion (tMCAO) were established for cerebral ischaemia-reperfusion injury. MCC950 was utilized as a selective NLRP3 inflammasome inhibitor. NLRP3 inflammasome associated protein, IL-23/IL-17 and IL-23R were detected to investigate their changes in the brain tissue after tMCAO. MCC950 inhibited the NLRP3 inflammasome, which alleviated the neurological ischaemia-reperfusion injury. Inhibition the NLRP3 inflammasome signalling by treatment with MCC950 decreased the activation of IL-23/IL-17 axis and the expression of IL-23R. The NLRP3 inflammasome facilitated the injury effect of the IL-23/IL-17 axis, which contributed to the cerebral ischaemia-reperfusion injury. This process was associated with IL-23R. Furthermore, this indicated that the NLRP3 inflammasome, as an important therapeutic target for ischaemic stroke, involves multiple mechanisms in ischaemia-reperfusion injury, and MCC950 is a promising way for clinical treatment.