Merck
  • Effect of Roasting and Storage on the Formation of Maillard Reaction and Sugar Degradation Products in Hazelnuts ( Corylus avellana L.).
Effect of Roasting and Storage on the Formation of Maillard Reaction and Sugar Degradation Products in Hazelnuts ( Corylus avellana L.).
Journal of agricultural and food chemistry (2018-12-12)
Neslihan Göncüoğlu Taş, Vural Gökmen
ABSTRACT
This study investigated the effect of roasting (150 °C for 30 min) and storage (12 months at 4 °C, 25 °C, and 25 °C in vacuum package), conditions of which are generally applied in the industry and markets, on the formation of Maillard reaction and sugar degradation products, namely dicarbonyl compounds, 5-hydroxymethylfurfural, N-ε-fructosyllysine, and N-ε-carboxymethyllysine in Tombul and Levant hazelnuts. Roasting increased all dicarbonyl compounds significantly ( p < 0.05). The concentration of methylglyoxal was the highest while 1-deoxyglucosone was the lowest in roasted hazelnuts. 5-Hydroxymethylfurfural and N-ε-carboxymethyllysine also increased significantly ( p < 0.05) with roasting while furosine decreased. Roasting changed the progress of the Maillard reaction from the early stage to the advanced stage. On the other hand, there were no significant changes ( p > 0.05) in the concentration of Maillard reaction and sugar degradation products independent of the storage conditions or time and hazelnut variety, except for glyoxal, diacetyl, and 1-deoxyglucosone. Therefore, neither 5-hydroxymethylfurfural nor furosine is suggested as a storage marker of the Maillard reaction and sugar degradation.