- Chiral allene-containing phosphines in asymmetric catalysis.
Chiral allene-containing phosphines in asymmetric catalysis.
Journal of the American Chemical Society (2011-10-07)
Feng Cai, Xiaotao Pu, Xiangbing Qi, Vincent Lynch, Akella Radha, Joseph M Ready
PMID21972824
ABSTRACT
We demonstrate that allenes, chiral 1,2-dienes, appended with basic functionality can serve as ligands for transition metals. We describe an allene-containing bisphosphine that, when coordinated to Rh(I), promotes the asymmetric addition of arylboronic acids to α-keto esters with high enantioselectivity. Solution and solid-state structural analysis reveals that one olefin of the allene can coordinate to transition metals, generating bi- and tridentate ligands.