Merck
  • Development and validation of a rapid and sensitive UPLC-MS/MS assay for simultaneous quantification of paclitaxel and cyclopamine in mouse whole blood and tissue samples.
Development and validation of a rapid and sensitive UPLC-MS/MS assay for simultaneous quantification of paclitaxel and cyclopamine in mouse whole blood and tissue samples.
Biomedical chromatography : BMC (2019-02-26)
Cheng-Hui Hsiao, Jun Zhao, Song Gao, Nashid Farhan, Yang Wang, Chun Li, Diana S-L Chow
ABSTRACT
The prominent stromal compartment surrounds pancreatic ductal adenocarcinoma and protects the tumor cells from chemo- or radiotherapy. We hypothesized that our nano formulation carrying cyclopamine (CPA, stroma modulator) and paclitaxel (PTX, antitumor agent) could increase the permeation of PTX through the stromal compartment and improve the intratumoral delivery of PTX. In the present study a sensitive, reliable UPLC-MS/MS method was developed and validated to quantify PTX and CPA simultaneously in mouse whole blood, pancreas, liver and spleen samples. Docetaxel was used as the internal standard. The method demonstrated a linear range of 0.5-2000 ng/mL for whole blood and tissue homogenates for both PTX and CPA. The accuracy and precision of the assay were all within ±15%. Matrix effects for both analytes were within 15%. Recoveries from whole blood, liver, spleen and pancreas homogenates were 92.7-105.2% for PTX and 72.8-99.7% for CPA. The stability was within ±15% in all test biomatrices. The validated method met the acceptance criteria according to US Food and Drug Administration regulatory guidelines. The method was successfully applied to support a pharmacokinetic and biodistribution study for PTX and CPA in mice biomatrices.