Merck
  • Rhamnocitrin isolated from Prunus padus var. seoulensis: A potent and selective reversible inhibitor of human monoamine oxidase A.
Rhamnocitrin isolated from Prunus padus var. seoulensis: A potent and selective reversible inhibitor of human monoamine oxidase A.
Bioorganic chemistry (2018-11-06)
Seung Cheol Baek, Mi Hyeon Park, Hyung Won Ryu, Jae Pil Lee, Myung-Gyun Kang, Daeui Park, Chul Min Park, Sei-Ryang Oh, Hoon Kim
ABSTRACT
Three flavanones and two flavones were isolated from the leaves of Prunus padus var. seoulensis by the activity-guided screening for new monoamine oxidase (MAO) inhibitors. Among the compounds isolated, rhamnocitrin (5) was found to potently and selectively inhibit human MAO-A (hMAO-A, IC50 = 0.051 µM) and effectively inhibit hMAO-B (IC50 = 2.97 µM). The IC50 value of 5 for hMAO-A was the lowest amongst all natural flavonoids reported to date, and the potency was 20.2 times higher than that of toloxatone (1.03 µM), a marketed drug. In addition, 5 reversibly and competitively inhibited hMAO-A and hMAO-B with Ki values of 0.030 and 0.91 µM, respectively. Genkwanin (4) was also observed to strongly inhibit hMAO-A and hMAO-B (IC50 = 0.14 and 0.35 µM, respectively), and competitively inhibit hMAO-A and hMAO-B (Ki = 0.097 and 0.12 µM, respectively). Molecular docking simulation reveals that the binding affinity of 5 with hMAO-A (-18.49 kcal/mol) is higher than that observed with hMAO-B (0.19 kcal/mol). Compound 5 interacts with hMAO-A at four possible residues (Asn181, Gln215, Thr336, and Tyr444), while hMAO-B forms a single hydrogen bond at Glu84. These findings suggest that compound 5 as well as 4 can be considered as novel potent and reversible hMAO-A and/or hMAO-B inhibitors or useful lead compounds for future development of hMAO inhibitors in neurological disorder therapies.