Merck
  • Nucleolin and heterogeneous nuclear ribonucleoprotein C proteins specifically interact with the 3'-untranslated region of amyloid protein precursor mRNA.
Nucleolin and heterogeneous nuclear ribonucleoprotein C proteins specifically interact with the 3'-untranslated region of amyloid protein precursor mRNA.
The Journal of biological chemistry (1995-07-21)
S H Zaidi, J S Malter
ABSTRACT
The central nervous system deposition by neurons and glia of beta A4 amyloid protein is an important contributing factor to the development of Alzheimer's disease. Amyloidogenic cells overexpress amyloid precursor protein (APP) mRNAs suggesting a transcriptional or post-transcriptional defect may contribute to this process. We have previously shown that APP mRNAs display regulated stability which is dependent on a 29-base element within the 3'-untranslated region (UTR). This domain specifically interacted with several cytoplasmic RNA-binding proteins. We have purified these APP RNA-binding proteins from a human T-cell leukemia and demonstrate that five cytoplasmic proteins of 70, 48, 47, 39, and 38 kDa form the previously observed APP RNA protein complexes. Amino acid sequence analyses showed that the 70-, 48-, and 47-kDa proteins were fragments of nucleolin and that the 39- and 38-kDa proteins were heterogeneous nuclear ribonucleoprotein (hnRNP) C protein. Northwestern and Western blot analyses of purified material further confirmed these data. Nucleolin protein is known to shuttle between the nucleus and cytoplasm but hnRNP C has not been reported within the cytoplasm. This report of sequence specific, mRNA binding by nucleolin and hnRNP C suggests that these proteins participate in the post-transcriptional regulation of APP mRNA through 3'-UTR, site-specific interactions.
Sigma-Aldrich
Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)