Merck
Axon regeneration: Vaccinating against spinal cord injury.
Current biology : CB (2000-02-19)
M T Filbin
ABSTRACT
Myelin is a potent inhibitor of axon regeneration, but has been viewed as just one of many factors that prevent regeneration after injury. So it comes as a surprise that immunization against myelin has been found to allow extensive axon regeneration after injury, without apparent autoimmune-induced demyelination.
Sigma-Aldrich
Neurite Outgrowth Assay Kit (3 µm), The NS220 Neurite Outgrowth Assay Kit (3 µm) is based on the use of Millicell cell culture inserts (chambers) containing a permeable membrane with 3-μm pores at the base.
Sigma-Aldrich
Neurite Outgrowth Assay Kit (1 µm), The NS225 Neurite Outgrowth Assay Kit (1 μm) is based on the use of Millicell cell culture inserts (chambers) containing a permeable membrane with 1 μm pores at the base.