Merck
  • The synthesis and localization of alternatively spliced fibronectin EIIIB in resting and thrombin-treated megakaryocytes.
The synthesis and localization of alternatively spliced fibronectin EIIIB in resting and thrombin-treated megakaryocytes.
Blood (1996-03-01)
P K Schick, C M Wojenski, V D Bennett, T Ivanova
ABSTRACT
There are several species of alternatively spliced fibronectin (FN). One of these, FN EIIIB, is primarily present in embryonic and in proliferating and migrating cells and is believed to be important for cell maturation. We have studied the synthesis, localization, and secretion of this FN isoform in isolated guinea pig megakaryocytes, nonmegakaryocytic bone marrow cells, and platelets. There was 7.5 times more general FN in megakaryocytes than in nonmegakaryocytic cells based on the analysis of equivalent amounts of protein. FN EIIIB was detected by Western blotting in megakaryocytes but not in nonmegakaryocytic cells present in bone marrow. Neither megakaryocytes nor platelets secreted FN EIIIB, while megakaryocytes secreted 25.3% +/- 4.6% general FN and platelets secreted about 61% general FN in response to thrombin. Analysis of immunostained cells by confocal microscopy revealed that FN EIIIB had been redistributed to the surface of megakaryocytes in response to thrombin. Synthesis was studied by metabolic labeling, and megakaryocytes were shown to synthesize FN and FN EIIIB. Thus, megakaryocytes and platelets are among a small number of adult cells and tissues that synthesize and contain FN EIIIB. The expression of FN EIIIB on the megakaryocyte surface may influence migration and maturation.
Product Description
Millipore
Protein A–Agarose, lyophilized powder