Merck
  • Specific and direct binding of protein kinase C to an immobilized tamoxifen analogue.
Specific and direct binding of protein kinase C to an immobilized tamoxifen analogue.
Cancer research (1988-07-01)
C A O'Brian, G M Housey, I B Weinstein
ABSTRACT
We have previously demonstrated that tamoxifen and related triphenylethylene compounds are potent inhibitors of protein kinase C (PKC). The present study demonstrates that PKC binds specifically and reversibly to the antiestrogen N-didesmethyltamoxifen when the drug is coupled to CNBr-activated agarose through its primary amine, in the absence of lipid and other cofactors of the enzyme. PKC did not bind to 4-hydroxytamoxifen, which had been immobilized on epoxy-activated Sepharose through its hydroxyl moiety. This shows that the binding of PKC to immobilized N-didesmethyltamoxifen was not merely due to hydrophobic interactions, since N-didesmethyltamoxifen and 4-hydroxytamoxifen have nearly identical hydrophobicities. These results demonstrate that PKC has specific triphenylethylene-binding sites, which may mediate the inhibition of PKC activity by these antiestrogens.
Sigma-Aldrich
Epoxy-activated−Sepharose 6B, lyophilized powder
Pricing and availability is not currently available.