Merck
  • In vitro and in vivo induction and activation of nNOS by LPS in oligodendrocytes.
In vitro and in vivo induction and activation of nNOS by LPS in oligodendrocytes.
Journal of neuroimmunology (2010-08-21)
S Y Yao, A Ljunggren-Rose, N Chandramohan, W O Whetsell, S Sriram
ABSTRACT
There are currently four known isoforms of nitric oxide synthase (NOS). Of these, neuronal NOS (nNOS) is known to be present exclusively in neurons, endothelial NOS (eNOS) in vascular endothelium, while the inducible form of NOS (iNOS) is known to be activated in oligodendrocytes, astrocytes and microglia. The fourth isoform, mitochondrial NOS (mtNOS), represents a post-translational modification of nNOS. Using western blotting and real time-PCR, we show induction and activation of nNOS following culture of oligodendrocyte progenitor cells (OPC) with lipopolysaccharide (LPS). Activation of nNOS results in accumulation of peroxynitrite and tyrosine nitration of proteins in oligodendrocytes resulting in reduced cell viability. Injection of LPS in vivo into the corpus callosum of rats leads to the development of extensive demyelination of the white matter tracts. Immunostaining of regions close to the injection site shows the presence of nNOS, but not iNOS, in oligodendrocytes. Neither iNOS nor nNOS was seen in astrocytes in areas of demyelination. These studies suggest that activation of nNOS in oligodendrocytes leads to oligodendrocyte injury resulting in demyelination.
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-O4 Antibody, clone 81, clone 81 (mAB O4), Chemicon®, from mouse
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®