Merck
  • Creation of a Novel Class of Potent and Selective MutT Homologue 1 (MTH1) Inhibitors Using Fragment-Based Screening and Structure-Based Drug Design.
Creation of a Novel Class of Potent and Selective MutT Homologue 1 (MTH1) Inhibitors Using Fragment-Based Screening and Structure-Based Drug Design.
Journal of medicinal chemistry (2018-02-28)
Fredrik Rahm, Jenny Viklund, Lionel Trésaugues, Manuel Ellermann, Anja Giese, Ulrika Ericsson, Rickard Forsblom, Tobias Ginman, Judith Günther, Kenth Hallberg, Johan Lindström, Lars Boukharta Persson, Camilla Silvander, Antoine Talagas, Laura Díaz-Sáez, Oleg Fedorov, Kilian V M Huber, Ioanna Panagakou, Paulina Siejka, Mátyás Gorjánácz, Marcus Bauser, Martin Andersson
ABSTRACT
Recent literature has both suggested and questioned MTH1 as a novel cancer target. BAY-707 was just published as a target validation small molecule probe for assessing the effects of pharmacological inhibition of MTH1 on tumor cell survival, both in vitro and in vivo. (1) In this report, we describe the medicinal chemistry program creating BAY-707, where fragment-based methods were used to develop a series of highly potent and selective MTH1 inhibitors. Using structure-based drug design and rational medicinal chemistry approaches, the potency was increased over 10,000 times from the fragment starting point while maintaining high ligand efficiency and drug-like properties.
Sigma-Aldrich
BAY-707, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
BAY-604, ≥97% (HPLC)
Pricing and availability is not currently available.