Merck
  • Dynamic changes of proteasome and protective effect of bortezomib, a proteasome inhibitor, in mice with acute pancreatitis.
Dynamic changes of proteasome and protective effect of bortezomib, a proteasome inhibitor, in mice with acute pancreatitis.
Biochemical and biophysical research communications (2018-09-22)
Qingtian Zhu, Xi Lin, Xinnong Liu, Tianyu Hou, Min Zhang, NingZhi Wang, Weiwei Chen, Guotao Lu, Weijuan Gong, Yanbing Ding, Weiming Xiao
ABSTRACT
The proteasome is involved in the activation of NF-κB and can regulate the progression of inflammatory diseases. However, the role of proteasome in acute pancreatitis (AP) has not been demonstrated. In this study, we first observed that the protein level and activity of proteasome 20S were increased significantly in pancreatic injury tissues after caerulein-induced mild acute pancreatitis (MAP) induction, which was in consistent with the expression of the NF-κB nucleoprotein and positively correlated with the severity of AP. Then, bortezomib, a classical proteasome inhibitor, was used to intervene the progression of MAP in mice. The results showed that bortezomib administration reduced the serum amylase and lipase levels and mitigated histopathological manifestation of pancreatic injury in mice. Meanwhile, bortezomib decreased the expression of NF-κB p65 nucleoprotein as well as total proteasome 20S protein, and inhibited the activity of 20S in pancreatic tissues. In addition, we found that bortezomib could protect pancreatic acinar cell against necrosis and mitigate the severity of AP in a severe acute pancreatitis model induced by sodium taurocholate hydrate. Taken together, our study for the first time confirmed that the proteasome participated in the pathogenesis of AP and its inhibitor bortezomib could protect against AP in mice.
Sigma-Aldrich
Proteasome 20S Activity Assay Kit, sufficient for 100 fluorometric tests
Pricing and availability is not currently available.