Merck
  • On the inhibition of capsaicin response in dorsal root ganglion neurons by nobilamide B and analogues: a structure-activity relationship study.
On the inhibition of capsaicin response in dorsal root ganglion neurons by nobilamide B and analogues: a structure-activity relationship study.
MedChemComm (2018-11-16)
Oliver John V Belleza, Jortan O Tun, Gisela P Concepcion, Aaron Joseph L Villaraza
ABSTRACT
Nobilamide B, a TRPV1 antagonist, and a series of Ala-substituted analogues were synthesized and their neuroactivity was assessed in a primary culture of dorsal root ganglion (DRG) neurons. Analogues 4, 6, and 7 showed comparable activity, affecting capsaicin response in neurons, in contrast to 2, 3, and 5 which showed minimal blocking. Compounds 2, 3, and 5 correspond to analogues in which d-Phe, d-Leu and d-allo-Thr have been substituted with Ala, respectively. The observed loss of bioactivity in these three analogues highlights the importance of d amino acids in the primary structure of nobilamide B.
Sigma-Aldrich
Fmoc-D-Phe-OH, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Pricing and availability is not currently available.