Merck
  • Pd-Catalyzed Carbonylative Carboperfluoroalkylation of Alkynes. Through-Space 13C-19F Coupling as a Probe for Configuration Assignment of Fluoroalkyl-Substituted Olefins.
Pd-Catalyzed Carbonylative Carboperfluoroalkylation of Alkynes. Through-Space 13C-19F Coupling as a Probe for Configuration Assignment of Fluoroalkyl-Substituted Olefins.
The Journal of organic chemistry (2017-07-06)
Sylwester Domański, Olga Staszewska-Krajewska, Wojciech Chaładaj
ABSTRACT
A four-component Pd-catalyzed protocol for direct synthesis of perfluoroalkyl-substituted enones is reported. Under mild conditions and low catalyst loading, alkynes, iodoperfluoroalkanes, (hetero)arylboronic acids, and carbon monoxide are assembled into highly elaborate products with good yields and excellent regio- and stereoselectivities. The configuration of the products was confirmed by the observation of through-space 13C-19F couplings, accessible through the analysis of routine 13C NMR spectra.
Sigma-Aldrich
(R)-Tol-BINAP Pd G3, ≥95%