Merck
  • Deformation Modes of Giant Unilamellar Vesicles Encapsulating Biopolymers.
Deformation Modes of Giant Unilamellar Vesicles Encapsulating Biopolymers.
ACS synthetic biology (2018-02-01)
Taiji Okano, Koya Inoue, Kaoru Koseki, Hiroaki Suzuki
ABSTRACT
The shapes of giant unilamellar vesicles (GUVs) enclosing polymer molecules at relatively high concentration, used as a model cytoplasm, significantly differ from those containing only small molecules. Here, we investigated the effects of the molecular weights and concentrations of polymers such as polyethylene glycol (PEG), bovine serum albumin (BSA), and DNA on the morphology of GUVs deflated by osmotic pressure. Although small PEG (MW < 1000) does not alter the mode of shape transformation even at >10% (w/w), PEG with MW > 6000 induces budding and pearling transformation at above 1% (w/w). Larger PEG frequently induced small buddings and tubulation from the membrane of mother GUVs. A similar trend was observed with BSA, indicating that the effect is irrelevant to the chemical nature of polymers. More surprisingly, long strands of DNA (>105 bp) enclosed in GUVs induced budding transformation at concentrations as low as 0.01-0.1% (w/w). We expect that this molecular size dependency arises mainly from the depletion volume effect. Our results showed that curving, budding, and tubulation of lipid membranes, which are ubiquitous in living cells, can result from simple cell-mimics consisting of the membrane and cytosolic macromolecules, but without specific shape-determining proteins.
Product Description
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand