Merck
  • YAP promotes osteogenesis and suppresses adipogenic differentiation by regulating β-catenin signaling.
YAP promotes osteogenesis and suppresses adipogenic differentiation by regulating β-catenin signaling.
Bone research (2018-06-07)
Jin-Xiu Pan, Lei Xiong, Kai Zhao, Peng Zeng, Bo Wang, Fu-Lei Tang, Dong Sun, Hao-Han Guo, Xiao Yang, Shun Cui, Wen-Fang Xia, Lin Mei, Wen-Cheng Xiong
ABSTRACT
YAP (yes-associated protein) is a transcriptional factor that is negatively regulated by Hippo pathway, a conserved pathway for the development and size control of multiple organs. The exact function of YAP in bone homeostasis remains controversial. Here we provide evidence for YAP's function in promoting osteogenesis, suppressing adipogenesis, and thus maintaining bone homeostasis. YAP is selectively expressed in osteoblast (OB)-lineage cells. Conditionally knocking out Yap in the OB lineage in mice reduces cell proliferation and OB differentiation and increases adipocyte formation, resulting in a trabecular bone loss. Mechanistically, YAP interacts with β-catenin and is necessary for maintenance of nuclear β-catenin level and Wnt/β-catenin signaling. Expression of β-catenin in YAP-deficient BMSCs (bone marrow stromal cells) diminishes the osteogenesis deficit. These results thus identify YAP-β-catenin as an important pathway for osteogenesis during adult bone remodeling and uncover a mechanism underlying YAP regulation of bone homeostasis.
Sigma-Aldrich
Monoclonal Anti-YAP1 antibody produced in mouse, clone 2F12, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid