Merck
  • Enrichment of cancer stem cells via β-catenin contributing to the tumorigenesis of hepatocellular carcinoma.
Enrichment of cancer stem cells via β-catenin contributing to the tumorigenesis of hepatocellular carcinoma.
BMC cancer (2018-08-05)
Harshul Pandit, Yan Li, Xuanyi Li, Weizhong Zhang, Suping Li, Robert C G Martin
ABSTRACT
Hepatocellular carcinoma (HCC) is among the deadliest cancers due to its heterogeneity, contributing to chemoresistance and recurrence. Cancer stem-like cells (CSCs) are suggested to play an important role in HCC tumorigenesis. This study investigates the role of Wnt/β-catenin pathway in CSC enrichment and the capabilities of these CSCs in tumor initiation in orthotopic immunocompetent mouse model. HCC-CSCs were enriched using established serum-free culture method. Wnt/β-catenin pathway activation and its components were analyzed by western blot and qRT-PCR. The role of β-catenin in enrichment of CSC spheroids was confirmed using siRNA interference. Tumorigenic capabilities were confirmed using orthotopic immunocompetent mouse model by injecting 2 × 106 Hepa1-6 CSC spheroids or control cells in upper left liver lobe. The serum-free cultured Hepa1-6 cells demonstrated self-renewal, spheroid formation, higher EpCAM expression, increased Hoechst-33342 efflux, and upregulated Wnt/β-catenin signaling. Wnt/β-catenin pathway upregulation was implicated with the downstream targets, i.e., c-MYC, Cyclin-D1, and LEF1. Also, we found that GSK-3β serine-9 phosphorylation increased in Hepa1-6 spheroids. Silencing β-catenin by siRNA reversed spheroid formation phenotype. Mice injected with Hepa1-6 CSC spheroids showed aggressive tumor initiation and growth compared with mice injected with control cells. Successfully induced Hepa1-6 spheroids were identified with CSC-like properties. Aberrant β-catenin upregulation mediated by GSK-3β was observed in the Hepa1-6 spheroids. The β-catenin mediated CSC enrichment in the induced spheroids possesses the capability of tumor initiation in immunocompetent mice. Our study suggests plausible cell dedifferentiation mediated by β-catenin contributes to CSC-initiated HCC tumor growth in vivo.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.