Merck
  • Organocatalyzed Photocontrolled Radical Polymerization of Semifluorinated (Meth)acrylates Driven by Visible Light.
Organocatalyzed Photocontrolled Radical Polymerization of Semifluorinated (Meth)acrylates Driven by Visible Light.
Angewandte Chemie (International ed. in English) (2017-11-15)
Honghong Gong, Yucheng Zhao, Xianwang Shen, Jun Lin, Mao Chen
ABSTRACT
Fluorinated polymers are important materials that are widely used in many areas. Herein, we report the development of a metal-free photocontrolled radical polymerization of semifluorinated (meth)acrylates with a new visible-light-absorbing organocatalyst. This method enabled the production of a variety of semifluorinated polymers with narrow molar-weight distributions from semifluorinated trithiocarbonates or perfluoroalkyl iodides. The high performance of "ON/OFF" control and chain-extension experiments further demonstrate the utility and reliability of this method. Furthermore, to streamline the preparation of semifluorinated polymers, a scalable continuous-flow approach has been developed. Given the broad interest in fluorinated materials and photopolymerization, we expect that this method will facilitate the development of advanced materials with unique properties.
Sigma-Aldrich
10-Phenylphenothiazine, ≥95%