Merck
  • Local tissue manipulation via a force- and pressure-controlled AFM micropipette for analysis of cellular processes.
Local tissue manipulation via a force- and pressure-controlled AFM micropipette for analysis of cellular processes.
Scientific reports (2018-04-14)
Phillip Roder, Carsten Hille
ABSTRACT
Local manipulation of complex tissues at the single-cell level is challenging and requires excellent sealing between the specimen and the micromanipulation device. Here, biological applications for a recently developed loading technique for a force- and pressure-controlled fluidic force microscope micropipette are described. This technique allows for the exact positioning and precise spatiotemporal control of liquid delivery. The feasibility of a local loading technique for tissue applications was investigated using two fluorescent dyes, with which local loading behaviour could be optically visualised. Thus, homogeneous intracellular distribution of CellTracker Red and accumulation of SYTO 9 Green within nuclei was realised in single cells of a tissue preparation. Subsequently, physiological micromanipulation experiments were performed. Salivary gland tissue was pre-incubated with the Ca2+-sensitive dye OGB-1. An intracellular Ca2+ rise was then initiated at the single-cell level by applying dopamine via micropipette. When pre-incubating tissue with the nitric oxide (NO)-sensitive dye DAF-FM, NO release and intercellular NO diffusion was observed after local application of the NO donor SNP. Finally, local micromanipulation of a well-defined area along irregularly shaped cell surfaces of complex biosystems was shown for the first time for the fluidic force microscope micropipette. Thus, this technique is a promising tool for the investigation of the spatiotemporal effects of locally applied substances in complex tissues.
Sigma-Aldrich
DAF-FM, ≥98% (HPLC)
Pricing and availability is not currently available.