Merck
  • Activation of G protein-coupled estrogen receptor signaling inhibits melanoma and improves response to immune checkpoint blockade.
Activation of G protein-coupled estrogen receptor signaling inhibits melanoma and improves response to immune checkpoint blockade.
eLife (2018-01-18)
Christopher A Natale, Jinyang Li, Junqian Zhang, Ankit Dahal, Tzvete Dentchev, Ben Z Stanger, Todd W Ridky
ABSTRACT
Female sex and history of prior pregnancies are associated with favorable melanoma outcomes. Here, we show that much of the melanoma protective effect likely results from estrogen signaling through the G protein-coupled estrogen receptor (GPER) on melanocytes. Selective GPER activation in primary melanocytes and melanoma cells induced long-term changes that maintained a more differentiated cell state as defined by increased expression of well-established melanocyte differentiation antigens, increased pigment production, decreased proliferative capacity, and decreased expression of the oncodriver and stem cell marker c-Myc. GPER signaling also rendered melanoma cells more vulnerable to immunotherapy. Systemically delivered GPER agonist was well tolerated, and cooperated with immune checkpoint blockade in melanoma-bearing mice to dramatically extend survival, with up to half of mice clearing their tumor. Complete responses were associated with immune memory that protected against tumor rechallenge. GPER may be a useful, pharmacologically accessible target for melanoma.
Sigma-Aldrich
Anti-GPER1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution