Merck
Increased formate overflow is a hallmark of oxidative cancer.
Nature communications (2018-04-11)
Johannes Meiser, Anne Schuster, Matthias Pietzke, Johan Vande Voorde, Dimitris Athineos, Kristell Oizel, Guillermo Burgos-Barragan, Niek Wit, Sandeep Dhayade, Jennifer P Morton, Emmanuel Dornier, David Sumpton, Gillian M Mackay, Karen Blyth, Ketan J Patel, Simone P Niclou, Alexei Vazquez
ABSTRACT
Formate overflow coupled to mitochondrial oxidative metabolism\ has been observed in cancer cell lines, but whether that takes place in the tumor microenvironment is not known. Here we report the observation of serine catabolism to formate in normal murine tissues, with a relative rate correlating with serine levels and the tissue oxidative state. Yet, serine catabolism to formate is increased in the transformed tissue of in vivo models of intestinal adenomas and mammary carcinomas. The increased serine catabolism to formate is associated with increased serum formate levels. Finally, we show that inhibition of formate production by genetic interference reduces cancer cell invasion and this phenotype can be rescued by exogenous formate. We conclude that increased formate overflow is a hallmark of oxidative cancers and that high formate levels promote invasion via a yet unknown mechanism.
Sigma-Aldrich
Anti-SHMT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-SHMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone V284, purified from hybridoma cell culture