Merck
  • Association of MMP-2, RB and PAI-1 with decreased recurrence-free survival and overall survival in bladder cancer patients.
Association of MMP-2, RB and PAI-1 with decreased recurrence-free survival and overall survival in bladder cancer patients.
Oncotarget (2017-12-17)
Owen T M Chan, Hideki Furuya, Ian Pagano, Yoshiko Shimizu, Kanani Hokutan, Lars Dyrskjøt, Jørgen Bjerggaard Jensen, Per-Uno Malmstrom, Ulrika Segersten, Filip Janku, Charles J Rosser
ABSTRACT
We previously reported an accurate urine-based bladder cancer (BCa)-associated diagnostic signature that can be used to non-invasively detect BCa. In this study, we investigated whether a component of this signature could risk stratify patients with BCa. Utilizing immunohistochemistry, we investigated angiogenin, MMP-2, p53, RB and PAI-1 expression from 939 patients with BCa. The expression levels were scored by assigning a proportion score and an intensity score to yield a total staining score for each protein. The expressions of each protein individually and as an aggregate were then correlated with progression-free survival (PFS), cancer-specific survival (CSS) and overall survival (OS). Differential expressions of these markers were noted in BCa. With multivariate analysis in non-muscle invasive bladder cancer (NMIBC) age, tumor grade portended a worse PFS, while age, tumor grade, nodal status, MMP2, RB and PAI-1 expression portended a worse OS. As for multivariate analysis in muscle invasive bladder cancer (MIBC), age MMP-2 and RB were associated with a worse PFS, while age, nodal status, MMP-2, RB and PAI-1 were associated with a worse OS. Using Kaplan-Meier survival analysis, we noted a significant reduction in OS as more of the five biomarkers were expressed in a tumor. Thus, overall, high expressions of MMP-2, RB and/or PAI-1 in bladder tumors were markers of poor prognosis. Individually, MMP-2, RB and PAI-1, as well as in aggregate correlated with poor survival in patients with BCa. Thus, patients whose bladder tumors express these biomarkers may benefit from early radical treatment and/or neoadjuvant or adjuvant therapies.
Sigma-Aldrich
Anti-SERPINE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.