![TSKgel® SW Type Guard Column phase diol, L × I.D. 7.5 cm × 21.5 mm, 13 μm particle size](/deepweb/assets/sigmaaldrich/product/images/327/408/ffe670b0-38ea-4aa4-8260-b0c490e62780/640/ffe670b0-38ea-4aa4-8260-b0c490e62780.jpg)
TSKgel® SW Type (13 µm) HPLC Column
![TSKgel® SW Type Guard Column phase diol, L × I.D. 7.5 cm × 21.5 mm, 13 μm particle size](/deepweb/assets/sigmaaldrich/product/images/327/408/ffe670b0-38ea-4aa4-8260-b0c490e62780/640/ffe670b0-38ea-4aa4-8260-b0c490e62780.jpg)
Synonym(s):
TSKgel® Size Exclusion (SW-Type) HPLC Guard Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing