
Trillin

Synonym(s):
Diosgenin glucoside, (25R)-3B-(β-D-Glucopyranosyloxy)spirost-5-ene, Alliumosid A, Collettiside I, Diosgenin β-D-glucoside, Disogluside, Funkioside A, Lilioglycoside A, Melongoside B, Polygonatoside A, Polyphyllin A, Prosapogenin D′3, Trillin
Empirical Formula (Hill Notation):
C33H52O8
CAS Number:
Molecular Weight:
576.76
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing