![Tricaprin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/210/692/6c3ee1d0-f70a-4400-9d6c-6130898f2b66/640/6c3ee1d0-f70a-4400-9d6c-6130898f2b66.png)
Tricaprin
![Tricaprin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/210/692/6c3ee1d0-f70a-4400-9d6c-6130898f2b66/640/6c3ee1d0-f70a-4400-9d6c-6130898f2b66.png)
Synonym(s):
Glyceryl tridecanoate, 1,2,3-Tricaprinoylglycerol, 1,2,3-Tridecanoylglycerol, Glycerol tricaprate, Glycerol tris(decanoate), Tricaprin, Tridecanoin
Linear Formula:
[CH3(CH2)8COOCH2]2CHOCO(CH2)8CH3
CAS Number:
Molecular Weight:
554.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing