Skip to Content
Merck
TOYOPEARL® AF-Tresyl-650M Bulk Media phase tresyl-activated, bottle of 5 g, 65 μm particle size

TOYOPEARL® AF-Tresyl-650M Bulk Media

TOYOPEARL® AF-Tresyl-650M Bulk Media phase tresyl-activated, bottle of 5 g, 65 μm particle size
Synonym(s):
TOYOPEARL® AF-Tresyl-650M Bulk Media

Products

Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1