![Tolnaftate for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/280/274/00750100-7d08-463a-b37c-c9ed1c756b53/640/00750100-7d08-463a-b37c-c9ed1c756b53.png)
Tolnaftate for system suitability
![Tolnaftate for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/280/274/00750100-7d08-463a-b37c-c9ed1c756b53/640/00750100-7d08-463a-b37c-c9ed1c756b53.png)
Synonym(s):
Tolnaftate, Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester
Empirical Formula (Hill Notation):
C19H17NOS
CAS Number:
Molecular Weight:
307.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing