Skip to Content
Merck
vibrant-m

Spectinomycin sulfate tetrahydrate

vibrant-m
Empirical Formula (Hill Notation):
C14H26N2O11S · 4H2O
CAS Number:
Molecular Weight:
502.49

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1