Sodium disulfite
Synonym(s):
Sodium disulfite
Empirical Formula (Hill Notation):
Na2O5S2
CAS Number:
Molecular Weight:
190.11
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2