Skip to Content
Merck
Retinyl Propionate Pharmaceutical Secondary Standard; Certified Reference Material

Retinyl propionate

Retinyl Propionate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Vitamin A propionate, Retinyl propionate
Empirical Formula (Hill Notation):
C23H34O2
CAS Number:
Molecular Weight:
342.51
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique