
Pseudomonas aeruginosa WDCM 00024 Vitroids™

Products
Brand
Feature
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing