Skip to Content
Merck
Propineb PESTANAL®, analytical standard

Propineb

Propineb PESTANAL®, analytical standard
Synonym(s):
Zinc propylenebis(dithiocarbamate)
Linear Formula:
[C5H8N2S4Zn]n
CAS Number:
EC Number:

Products

Product Category
Brand
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing