![Prednisolone acetate for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/289/103/c05b7404-3af2-4425-9b4a-708c9af4319c/640/c05b7404-3af2-4425-9b4a-708c9af4319c.png)
Prednisolone acetate for peak identification
![Prednisolone acetate for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/289/103/c05b7404-3af2-4425-9b4a-708c9af4319c/640/c05b7404-3af2-4425-9b4a-708c9af4319c.png)
Synonym(s):
Prednisolone 21-acetate, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione
Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing