![Prealbumin from human plasma lyophilized powder](/deepweb/assets/sigmaaldrich/product/images/291/295/c01a99f1-ad7d-46be-bf04-e40f802226fb/640/c01a99f1-ad7d-46be-bf04-e40f802226fb.jpg)
Prealbumin from human plasma
![Prealbumin from human plasma lyophilized powder](/deepweb/assets/sigmaaldrich/product/images/291/295/c01a99f1-ad7d-46be-bf04-e40f802226fb/640/c01a99f1-ad7d-46be-bf04-e40f802226fb.jpg)
Synonym(s):
Thyroxine binding prealbumin, Transthyretin
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing