Potassium Isoquinoline-4-trifluoroborate
Synonym(s):
Potassium 4-isoquinolinetrifluoroborate, Potassium isoquinoline-4-trifluoroborate
Empirical Formula (Hill Notation):
C9H6BF3KN
CAS Number:
Molecular Weight:
235.06
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing