PMSF
Synonym(s):
Phenylmethanesulfonyl fluoride, α-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, PMSF, Phenylmethylsulfonyl fluoride
Empirical Formula (Hill Notation):
C7H7FO2S
CAS Number:
Molecular Weight:
174.19
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing