Skip to Content
Merck
vibrant-m

Piretanide

vibrant-m
Synonym(s):
4-Phenoxy-3-(1-pyrrolidinyl)-5-sulfamoylbenzoic acid
Empirical Formula (Hill Notation):
C17H18N2O5S
CAS Number:
Molecular Weight:
362.40

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing