![Phenylephrine hydrochloride for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/339/180/7f442540-b275-47d9-a956-6fb8f51773b9/640/7f442540-b275-47d9-a956-6fb8f51773b9.png)
Phenylephrine hydrochloride for peak identification
![Phenylephrine hydrochloride for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/339/180/7f442540-b275-47d9-a956-6fb8f51773b9/640/7f442540-b275-47d9-a956-6fb8f51773b9.png)
Synonym(s):
(R)-(−)-Phenylephrine hydrochloride, (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride
Linear Formula:
HOC6H4CH(CH2NHCH3)OH·HCl
CAS Number:
Molecular Weight:
203.67
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing