![Panaxadiol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/916/991/3dba6bd9-2a59-407b-abf3-5170b7adfacc/640/3dba6bd9-2a59-407b-abf3-5170b7adfacc.png)
Panaxadiol
![Panaxadiol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/916/991/3dba6bd9-2a59-407b-abf3-5170b7adfacc/640/3dba6bd9-2a59-407b-abf3-5170b7adfacc.png)
Synonym(s):
(3β,12β,20R)-20,25-Epoxydammarane-3,12-diol
Empirical Formula (Hill Notation):
C30H52O3
CAS Number:
Molecular Weight:
460.73
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing