Oxaliplatin Related Compound A
Synonym(s):
Oxalic acid dihydrate, Ethanedioic acid dihydrate
Linear Formula:
HO2CCO2H · 2H2O
CAS Number:
Molecular Weight:
126.07
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2